Learn More
Gilson™ PIPETMANTM L Multichannel Pipette Trade-In PROMO
GREENER_CHOICE
Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.
Supplier:  Gilson™ FA10010T
 View more versions of this product
Catalog No. FA10010TG
Only null left
Description
Description
  • Gilson-patented volume locking system guarantees no accidental change in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • Redesigned handle fits naturally in your hand, regardless of the size
  • Unique internal design applies even, equal pressure from either side of the pipettes, rather than on the center of the channels like many other multichannel pipettes do
  • 2D identification code tracks product reference number, date of manufacture, serial number, and maximum volume of pipette
  • Available in 8- or 12- channels, in volumes ranging from 0.50 to 300 μL
Specifications
Chemical Identifiers
No
Manual
Ergonomic
Pipetman
Non-sterile
Ergonomic
5.98 in
Autoclavable
Reusable
Polypropylene
No
Pipette
1
Specifications
PIPETMAN DIAMOND Tips DL10, D200, DFL10ST, DF30ST
Autoclavable
Manual
Pipette Tip 0.5 to 20 μL
Polypropylene
12
No
Pipette
Ergonomic
5.98 in
No
White
Reusable
Ergonomic
P12x20L
Pipetman
Non-sterile
2 to 20 μL
1
Certified Quality
Product Suggestions
Product Suggestions