missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMANTM L Mechanical Pipette Trade-In PROMO

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

Supplier:  Gilson™ FA10007T

 View more versions of this product

Catalog No. FA10007TG


Only null left
Explore available promotions

Description

Description

  • Redesigned handle fits naturally in user's hand, regardless of size
  • Volume locking system guarantees no accidental changes in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • 2D identification code allows for easy tracking of product's information
  • New piston assembly designed to drastically improve pipetting and purge forces
  • Redesigned tip ejector shape and internal mechanism reduces tip ejection forces up to 50%
Specifications

Specifications

±12, ±15, ±30 μL
No
Purple
Reusable
Ergonomic
P5000L
Pipetman
Non-sterile
500 to 5000 μL
Transporting a measured volume of liquid
4.25 in
No Tip Ejector
Autoclavable
Manual
Pipette Tip 500 to 5000 μL
Polypropylene
1
No
Mechanical Pipette
Ergonomic
1
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions