Learn More
Gilson™ PIPETMANTM L Mechanical Pipette Trade-In PROMO
Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.
Supplier:  Gilson™ FA10006MT
 View more versions of this product
Catalog No. FA10006MTG
Only null left
Explore available promotions
Description
Description
  • Redesigned handle fits naturally in user's hand, regardless of size
  • Volume locking system guarantees no accidental changes in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • 2D identification code allows for easy tracking of product's information
  • New piston assembly designed to drastically improve pipetting and purge forces
  • Redesigned tip ejector shape and internal mechanism reduces tip ejection forces up to 50%
Specifications
Specifications
±3, ±4, ±8 μL
No
Blue
Reusable
Ergonomic
P1000L
Pipetman
Non-sterile
100 to 1000 μL
Transporting a measured volume of liquid
1
Certified Quality
Stainless Steel Ejector
Autoclavable
Manual
D-1000, D-1200, DF1000ST, DF1200ST Tip
Polypropylene
1
No
Mechanical Pipette
Ergonomic
D1000, D1200, DF1000ST, DF1200ST tips
4.25 in
Product Suggestions
Product Suggestions