Learn More
Gilson™ PIPETMANTM Multichannel Pipette
GREENER_CHOICE
The PIPETMANTM Multichannel Pipette reduces the force needed to aspirate and dispense liquids.
Supplier:  Gilson™ F144074
 View more versions of this product
Catalog No. F144074G
Only null left
Description
Description
  • PIPETMAN pipettes bring a new level of comfort into the Gilson range
  • Redesigned piston mechanism reduces the force needed to aspirate and dispense liquids
  • Offers one of the lowest pipetting forces commercially available
  • Maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes
  • New piston assembly system and high quality piston grease guarantee hours of comfortable pipetting
Specifications
Chemical Identifiers
Diamond Tips
No
Manual
Ergonomic
Pipetman
Non-sterile
1
12.87 in.
Certified Quality
Adjustable Volume
Autoclavable
Reusable
Metal Ejector
No
Manual Air Displacement Pipette
3 Years
32.7 cm
Specifications
Diamond Tips
No
Green
Reusable
Ergonomic
P8x300
Pipetman
Non-sterile
30 to 300 μL
3 Years
5.98 in.
12.87 in.
1.54 in.
.88 lb
Adjustable Volume
Autoclavable
Manual
Pipette Tip 50 to 1000 μL
Metal Ejector
8
No
Manual Air Displacement Pipette
1
330 x 152 x 38 mm
15.2 cm
32.7 cm
3.9 cm
Certified Quality
Product Suggestions
Product Suggestions