Learn More
Gilson™ PIPETMANTM Multichannel Pipette
GREENER_CHOICE
The PIPETMANTM Multichannel Pipette reduces the force needed to aspirate and dispense liquids.
Supplier:  Gilson™ F144071
 View more versions of this product
Catalog No. F144071G
Only null left
Description
Description
  • PIPETMAN pipettes bring a new level of comfort into the Gilson range
  • Redesigned piston mechanism reduces the force needed to aspirate and dispense liquids
  • Offers one of the lowest pipetting forces commercially available
  • Maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes
  • New piston assembly system and high quality piston grease guarantee hours of comfortable pipetting
Specifications
Chemical Identifiers
Diamond Tips
No
Manual
Ergonomic
Pipetman
Non-sterile
1
12.87 in.
Certified Quality
Adjustable Volume
Autoclavable
Reusable
Metal Ejector
No
Manual Air Displacement Pipette
3 Years
32.7 cm
Specifications
Diamond Tips
No
Yellow
Reusable
Ergonomic
P12x20
Pipetman
Non-sterile
2 to 20 μL
3 Years
6.02 in.
12.87 in.
1.5 in.
.88 lb
Adjustable Volume
Autoclavable
Manual
Pipette Tip 2 to 200 μL
Metal Ejector
12
No
Manual Air Displacement Pipette
1
330 x 152 x 38 mm
15.3 cm
32.7 cm
3.8 cm
Certified Quality
Product Suggestions
Product Suggestions