missing translation for 'onlineSavingsMsg'
Learn More

PIPES, disodium salt, 99%

CAS: 76836-02-7 | C8H16N2Na2O6S2 | 346.324 g/mol

$131.57 - $715.38

Chemical Identifiers

CAS 76836-02-7
Molecular Formula C8H16N2Na2O6S2
Molecular Weight (g/mol) 346.324
InChI Key GMHSTJRPSVFLMT-UHFFFAOYSA-L
Synonym 1, 4-Piperazinediethanesulfonic acid disodium salt
PubChem CID 173553
ChEBI CHEBI:63055
IUPAC Name disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215090250
SDS View Documents
Thermo Scientific Chemicals
215090250
25 g Glass bottle
Each for $131.57
Only null left
 
AC215091000
SDS View Documents
Thermo Scientific Chemicals
215091000
100 g Glass bottle
Each for $307.33
Only null left
 
AC215092500
SDS View Documents
Thermo Scientific Chemicals
215092500
250 g Glass bottle
Each for $715.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

76836-02-7
346.324
1, 4-Piperazinediethanesulfonic acid disodium salt
CHEBI:63055
C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+]
C8H16N2Na2O6S2
GMHSTJRPSVFLMT-UHFFFAOYSA-L
173553
disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate

Specifications

0.050 max. (0.1M in water) at 260nm, 0.050 max. (0.1M in water) at 280nm, 0.050 max. (260nm, 0.1M in water)
White
Crystalline Powder or Powder
99%
C8H16N2Na2O6S2
1, 4-Piperazinediethanesulfonic acid disodium salt
C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+]
346.324
CHEBI:63055
99%
76836-02-7
9.2 to 10.0 max. (25°C, 1% min. aq. soln.)
25 g
Authentic
Glass bottle
GMHSTJRPSVFLMT-UHFFFAOYSA-L
disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
173553
346.33
PIPES, disodium salt
Videos
SDS
Documents

Documents

RUO – Research Use Only