missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 98.5+%, for biochemistry

Catalog No. AC172610250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172610250 25 g Plastic bottle
AC172611000 100 g Plastic bottle
AC172615000 500 g Plastic bottle
AC172610025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC172610250 Supplier Thermo Scientific Chemicals Supplier No. 172610250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym 1, 4-Piperazinebis(ethanesulfonic acid)
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Absorbance (0.1M in 0.1M NaOH) (1 cm cell), (0.1M in 0.1M NaOH) (1 cm cell) at 0nm, 0.04 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M in 0.1M NaOH) (1cm cell)
Color White
Physical Form Micro-crystalline Powder
Quantity 25 g
Assay Percent Range 98.5+%
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Packaging Plastic bottle
Formula Weight 302.35
Percent Purity ≥98.50%
Grade Biochemical
Chemical Name or Material PIPES
Show More Show Less

RUO – Research Use Only