missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 1.0M buffer soln., pH 8.0

Catalog No. p-7041889
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60618AK 250 mL
AAJ60618AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60618AK Supplier Thermo Scientific Chemicals Supplier No. J60618AK
Only null left

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES was used in the preparation of a variety of marine and mammalian tissues for electron microscopic study. Lipid loss and artifacts are minimized when PIPES was used to buffer the glutaraldehyde fixative. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer. Alkaline phosphatase retention activity was 60% greater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES was used in the preparation of a variety of marine and mammalian tissues for electron microscopic study. Lipid loss and artifacts are minimized when PIPES was used to buffer the glutaraldehyde fixative. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer. Alkaline phosphatase retention activity was 60% greater.

Solubility
Fully misicible in water. Soluble in water (0.05 M), giving a clear, colorless solution (the resulting solution has a pH of approximately 4.5-4.7). 10 g will dissolve in 40 mL of 0.1 N NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, reducing agents, heat.

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Color Colorless
Concentration 1.0 M
pH 8.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully misicible in water. Soluble in water (0.05 M),giving a clear,colorless solution (the resulting solution has a pH of approximately 4.5-4.7). 10 g will dissolve in 40 mL of 0.1 N NaOH.
Chemical Name or Material PIPES buffer soln.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only