missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 0.5M buffer soln., pH 8.0

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

Supplier:  Thermo Scientific Chemicals J61406AK

 View more versions of this product

Catalog No. AAJ61406AK


Only null left

Description

Description

PIPES can be used in a wide array of biological applications, including as a component in media formulations. Specific applications include buffers for cell culture and diagnostic reagent production. It is used to buffer glutaraldehyde fixative solutions when fixing plant and/or animal tissue tissue samples. This will help prevent the loss of lipids. It is used to buffer glutaraldehyde and formaldehyde when fixing fungal zoospores for fluorescence microscopy and electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES can be used in a wide array of biological applications, including as a component in media formulations. Specific applications include buffers for cell culture and diagnostic reagent production. It is used to buffer glutaraldehyde fixative solutions when fixing plant and/or animal tissue tissue samples. This will help prevent the loss of lipids. It is used to buffer glutaraldehyde and formaldehyde when fixing fungal zoospores for fluorescence microscopy and electron microscopy.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents, bases and heat.
Specifications

Specifications

PIPES buffer soln.
Colorless
8.0
250 mL
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
79723
5625-37-6
0.5 M
Liquid
C8H18N2O6S2
Fully miscible in water.
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
302.36
CHEBI:44933

Chemical Identifiers

5625-37-6
302.36
IHPYMWDTONKSCO-UHFFFAOYSA-N
CHEBI:44933
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
C8H18N2O6S2
MFCD00006159
79723
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only