missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 0.5M buffer soln., pH 7.5

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

Supplier:  Thermo Scientific Chemicals J63617AK

 View more versions of this product

Catalog No. AAJ63617AK


Only null left

Description

Description

PIPES is frequently used as a buffering agent in biochemistry. PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES has been documented minimizing lipid loss when buffering glutaraldehyde histology in plant and animal tissues and has a negligible capacity to bind divalent ions. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer is done.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES is frequently used as a buffering agent in biochemistry. PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES has been documented minimizing lipid loss when buffering glutaraldehyde histology in plant and animal tissues and has a negligible capacity to bind divalent ions. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer is done.

Solubility
Soluble in 1M NaOH (0.5 M at 20°C), water (∼1 g/L at 100°C), and 0.5 N NaOH (0.1 M).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

PIPES buffer soln.
Colorless
7.5
250 mL
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
79723
5625-37-6
0.5 M
Liquid
C8H18N2O6S2
Soluble in 1M NaOH (0.5 M at 20°C),water (∽1g/L at 100°C),and 0.5 N NaOH (0.1 M).
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
302.36
CHEBI:44933

Chemical Identifiers

5625-37-6
302.36
IHPYMWDTONKSCO-UHFFFAOYSA-N
CHEBI:44933
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
C8H18N2O6S2
MFCD00006159
79723
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only