missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 0.5M buffer soln., pH 6.8

Catalog No. p-7042226
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61786AE 100 mL
AAJ61786AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61786AE Supplier Thermo Scientific Chemicals Supplier No. J61786AE
Only null left

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
MDL Number MFCD00006159
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Color Colorless
Concentration 0.5 M
pH 6.8
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material PIPES buffer soln.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only