missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Piperacillin Sodium Salt, MP Biomedicals™
Manufacturer: Mp Biomedicals 0215626805
Specifications
Piperacillin Sodium Salt | |
C23H26N5NaO7S | |
Pipracil, Sodium Piperacillin | |
[Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O | |
539.54 | |
CHEBI:8233 | |
Crystalline Powder | |
5g |
59703-84-3 | |
MFCD00917471 | |
WCMIIGXFCMNQDS-QMJSIVKPSA-M | |
sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
23666879 | |
539.5 | |
White |
Chemical Identifiers
59703-84-3 | |
539.54 | |
WCMIIGXFCMNQDS-QMJSIVKPSA-M | |
23666879 | |
sodium (2S,5R,6R)-6-[(2S)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
C23H26N5NaO7S | |
MFCD00917471 | |
Pipracil, Sodium Piperacillin | |
CHEBI:8233 | |
[Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O |
Safety and Handling
Recommended Storage : Store at 4°C.