missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PIP2:PS Lipid Kinase Substrate

Catalog No. PV5100
Encompass
Change view
Click to view available options
Quantity:
400 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5100 400 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5100 Supplier Thermo Scientific™ Supplier No. PV5100
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifically optimized for use with the Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), this substrate consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphotidylserine (PS).

Specifically optimized for use with the Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), this substrate consists of lipid vesicles of a 1:19 molar ratio of phosphoinositol-4,5-bisphosphate (PIP2) with phosphotidylserine (PS).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

Specifications

Assay Entry Biochemical ADP accumulation
Chromagen Quantity 400nmol
Content And Storage PIP2:PS Lipid Kinase Substrate in 50 mM HEPES pH 7.5, 3 mM MgCl2, and 1 mM EGTA.

Store at -20°C.
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, protein assays and analysis, target and lead identification and validation, Class I PI3 kinases (PIK3CA, PIK3CB, PIK3CD, and PIK3CG), Adapta™ Universal Kinase Assay (PV5099), drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity
Quantity 400 nmol
Substrate Phosphoinositol-4,5-bisphosphate (PIP2) lipid vesicles with phosphotidylserine (PS)
Substrate Properties Lipid-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Lipid Kinases
Gene ID (Entrez) 5290, 5294
Product Type PIP2:PS Lipid Kinase Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Target Entry PIK3CG, PIK3CD/PIK3R1, PIK3CA/PIK3R1
Validated Kinases PIK3CA, PIK3CG, PIK3CB, PIK3CD
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.