missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pioglitazone Hydrochloride 98.0+%, TCI America™
$313.28 - $937.92
Chemical Identifiers
| CAS | 112529-15-4 |
|---|---|
| Molecular Formula | C19H21ClN2O3S |
| Molecular Weight (g/mol) | 392.898 |
| MDL Number | MFCD04975446 |
| InChI Key | GHUUBYQTCDQWRA-UHFFFAOYSA-N |
| Synonym | 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione Hydrochloride |
| PubChem CID | 60560 |
| IUPAC Name | 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride |
| SMILES | CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl |
Chemical Identifiers
| 112529-15-4 | |
| 392.898 | |
| GHUUBYQTCDQWRA-UHFFFAOYSA-N | |
| 60560 | |
| CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl |
| C19H21ClN2O3S | |
| MFCD04975446 | |
| 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione Hydrochloride | |
| 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride |
Specifications
| 112529-15-4 | |
| C19H21ClN2O3S | |
| White | |
| 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione Hydrochloride | |
| CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl | |
| 392.898 | |
| 60560 | |
| ≥98.0% (HPLC,N) |
| Powder/Crystals | |
| MFCD04975446 | |
| 191°C | |
| GHUUBYQTCDQWRA-UHFFFAOYSA-N | |
| 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride | |
| 5 g | |
| 392.90 | |
| Pioglitazone Hydrochloride |
Safety and Handling
TSCA : No