missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Pierceable Stoppers for Vacuum Blood Tubes and Test Tubes

For recapping vacuum blood tubes and test tubes and can easily be pierced with syringe.

Supplier:  Globe Scientific 113150B

 View more versions of this product

Catalog No. 22-010-027


Only null left
Explore available promotions

Description

Description

  • Material: Thermoplastic Elastomer (TPE)
  • Temperature Range: −80°C to +121°C
  • Superior leak-resistance
  • Latex free
  • Non-sterile
  • Autoclavable
Specifications

Specifications

Pierceable Stopper
Autoclavable
Blue
Non-sterile
13 mm
TPE
1000
Vacuum Blood Tubes and Test Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions