missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast DNA Extraction Kit

Catalog No. PI78870
Change view
Click to view available options
Quantity:
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78870 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI78870 Supplier Thermo Scientific™ Supplier No. 78870

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quickly, efficiently and easily lyse yeast cells to release DNA with Y-PER lysis reagent.

Quickly and easily lyse yeast cells to release DNA for study with Thermo Scientific™ Pierce Yeast DNA Extraction Kit, which utilizes a Thermo Scientific γ-PER Reagent.

The kit provides greater yields than traditional methods of DNA isolation from yeast. Typically, extraction and purification of DNA from yeast are time-consuming and labor-intensive. The yeast proteinaceous cell wall is notoriously difficult to lyse and requires harsh treatments that are time-consuming and can damage the extracted DNA. The Yeast DNA Extraction Kit protocol requires less than one hour to complete, is effective without enzymatic treatment or glass beads, and yields little to no RNA contamination regardless of RNase presence. In studies with Saccharomyces cerevisiae, high yields of genomic and plasmid DNA are consistently obtained. DNA purified using this kit is suitable for PCR amplification, bacterial transformations, restriction digestions and hybridization applications.

Highlights:

  • Eliminates need for glass beads or harsh enzyme treatments
  • Yields DNA that is compatible with polymerase chain reaction (PCR) amplification
  • Rapidly isolates plasmid DNA from S. cerevisiae suitable for transformation of E. coli
  • Protocol is easily scalable for use with single colonies or 500mL cultures

Includes:

Thermo Scientific γ-PER Yeast Protein Extraction Reagent, DNA releasing reagents A and B, and protein removal reagent

Specifications

Content And Storage Sufficient For: 50 purifications from 10 mL cultures
• Y-PER Yeast Protein Extraction Reagent, 25 mL
• DNA Releasing Agent A, 20 mL
• DNA Releasing Agent B, 20 mL
• Protein Removal Reagent, 10 mL

Store at 4°C. Precipitates may form; gently warm at 37°C for 1–5 minutes.
Sample Type Yeast
For Use With (Application) PCR, Sequencing, Transformation
High-throughput Compatibility Not High-throughput Compatible
Product Type Yeast DNA Extraction Kit
Quantity 75 mL
Target Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.