missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Urea

Catalog No. PI29700
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI29700 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI29700 Supplier Thermo Scientific™ Supplier No. 29700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pure, crystalline urea (CH4N2O), CAS 57-13-6; denaturant with low UV-absorptivity for protein biology methods (elution, solubilization, refolding, storage).

Thermo Scientific™ Pierce Urea is pure, crystalline urea, a protein denaturant with low UV-absorptivity.

Additional Information and Online Ordering

Highlights:

  • Melting point: 132-136°C
  • Specification: A280 <0.100

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Form Powder
Reagent Type Chaotrope, Denaturant
Quantity 1 kg
Product Type Urea

For Research Use Only. Not for use in diagnostic procedures.