missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis

Catalog No. PI88702
Change view
Click to view available options
Quantity:
100 kU
25 kU
5 kU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88702 100 kU
PI88700 5 kU
PI88701 25 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI88702 Supplier Thermo Scientific™ Supplier No. 88702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.

Thermo Scientific Pierce Universal Nuclease for Cell Lysis is ideal for a wide variety of applications where complete digestion of nucleic acids is needed when preparing cell lysates.

Features of Universal Nuclease for Cell Lysis:

  • Broad spectrum—degrades all forms of DNA and RNA
  • Highest-quality enzyme—nuclease is ≥99% pure, as tested by SDS-PAGE
  • Robust activity—100-fold greater specific activity than DNase I
  • Versatile—can be used with a wide variety of cell lysis reagents

Pierce Universal Nuclease for Cell Lysis is a genetically engineered endonuclease from Serratia marcescens. The enzyme is produced and purified from E. coli and consists of two identical 30-kDa subunits with two critical disulfide bonds. This indiscriminate endonuclease degrades single-stranded, double-stranded, linear and circular DNA and RNA and is effective over a wide range of temperatures and pH. This enzyme has high specific activity (100-fold greater than DNase I) and increased thermal stability compared to other nucleases. Pierce Universal Nuclease is ≥99 pure enzyme, is free of any measurable protease activity and is supplied at 250 U/μL. Pierce Universal Nuclease for Cell Lysis is identical in performance to Benzonase™ Nuclease (EMD Merck).

Applications:

  • Use with B-PER, Y-PER or other commercial or homebrew cell lysis reagents and/or mechanical disruption to reduce viscosity in protein extracts
  • Remove DNA and RNA from recombinant protein preparations prior to downstream processing

Pierce Universal Nuclease for Cell Lysis is commonly used to reduce the viscosity of bacterial and mammalian protein extracts for downstream application by removing the nucleic acids from protein preparations. The enzyme completely digests nucleic acids to oligonucleotides that are less than 5 bases long. Pierce Universal Nuclease for Cell Lysis helps to improve the separation of the lysate pellet from the supernatant, enhances filtration of the treated lysate, improves chromatography processing time and increases the overall protein yield. The endonuclease has also been shown to improve the compatibility of protein extracts for 2D gel electrophoresis. One unit corresponds to the amount of enzyme required to produce a change of 1.0 in the absorbance at 260 nm of sonicated Herring DNA over 30 minutes at 37°C, as determined using standard nuclease from the Merck™ Serratia marcescens volumetric activity assay.

Related Products

Micrococcal Nuclease Solution (≥ 1 unit/μL) (Cat. No. 88216)

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Store in a cool, dry, well-ventilated area, protected from direct sunlight.
Form Liquid
Product Line Pierce
Reagent Type Enzyme for Cell Lysis
Enzyme Nuclease
Quantity 100 kU
After cell lysis, my mass spectrometry sample is very viscous and difficult to pipette. How do I reduce the sample viscosity?

High sample viscosity after lysis is due to release of DNA from the nucleus. Sonication or addition of a nuclease such as the Pierce Universal Nuclease (Cat. No. 88700, 88701, or 88702) can be used to degrade DNA and reduce sample viscosity.

Can you explain how the B-PER Bacterial Protein Extraction Reagent lyses cells?

The B-PER Reagent solution contains a proprietary, mild, non-ionic detergent in 20 mM Tris-HCl, pH 7.5. It effectively disrupts cells and solubilizes native or recombinant proteins without denaturation. The reagent creates holes in the cell membrane that will leak out cytosolic proteins. The sample may become very viscous when the bacterial chromosome is released. We recommend adding DNAse I (Cat. No. 90083) to the reagent to reduce viscosity. For better lysis efficiency and if there are inclusion bodies, we recommend adding Lysozyme (Cat. No. 90082) to the reagent. Alternatively, you may purchase the B-PER Bacterial Protein Extraction Reagent with Enzymes Kit (Cat. No. 90078 or 90079) that includes the B-PER Bacterial Protein Extraction Reagent, DNase I, and Lysozyme.

For Research Use Only. Not for use in diagnostic procedures.