missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Ubiquitin Enrichment Kit

Catalog No. PI89899
Change view
Click to view available options
Quantity:
15 reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89899 15 reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89899 Supplier Thermo Scientific™ Supplier No. 89899

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Purify (enrich) poly-ubiquitinated proteins from cell and tissue lysates for post-translational modification studies using this affinity-resin, spin-column kit.

Thermo Scientific™ Pierce Ubiquitin Enrichment Kit facilitates the isolation of polyubiquitin protein conjugates from cultured cells and tissue samples.

The kit uses anti-ubiquitin affinity resin to bind and purify polyubiquitinated proteins (i.e., those with modifications containing more than four ubiquitin monomers) from cell lysates and other protein samples. The enriched fraction can subsequently be analyzed to determine the amount of general ubiquitin conjugates present or to identify a specific protein of interest by Western blotting. The assay protocol is fast and straight-forward, allowing for isolation of polyubiquitinated proteins and the fractionation of monoubiquitinated species in the resin flow-through. The Ubiquitin Enrichment Kit outperforms kits from other suppliers and provides a clean and specific preparation of proteins when compared to a control resin.

Highlights:

  • Fast – less than 45 minutes hands-on time

  • Complete – includes all reagents needed for ubiquitin-modified protein enrichment from cultured cells and tissue samples, as well as anti-ubiquitin antibody serum for Western blot analysis

  • Flexible – sample incubation from two hours to overnight allows assay to be completed in several hours or in less than 30 minutes after overnight incubation

  • Robust – compatible with all Thermo Scientific Pierce Cell Lysis Products and standard RIPA formulations

  • Multiple-sample format – easily processes 1 to 15 samples concurrently

Includes:

Polyubiquitin affinity resin (300μL), Tris-buffered saline pack, microcentrifuge spin columns and accessories, polyubiquitin positive control and anti-ubiquitin rabbit anti-serum

Specifications

Content And Storage Sufficient For: Enriching polyubiquitinated proteins from 15 lysate samples, each containing 0.15 mg total protein
• Polyubiquitin Positive Control (1000X), 50 μL (store at -20°C)
• Anti-ubiquitin Antibody (rabbit antiserum), 5 μL (store at -20°C)
• Polyubiquitin Affinity Resin Slurry, 300 μL (store at 4°C—do not freeze)
• TBS Pack (makes 500mL), 1 pack (store at 4°C)
• Spin Columns and Accessories, 18 columns (store at 4°C)
Product Line Pierce
Quantity 15 reactions kit
Target Polyubiquitinated Protein
Type Ubiquitin Enrichment Kit
Stationary Phase Proprietary Ligand
Sufficient For 15 Reactions
Format Kit, Microcentrifuge, Spin Column
Column Type Affinity, Agarose Resin
Can I isolate my proteins based on their post-translational modification (PTM) or active binding site?

Enrichment of specific proteins or protein complexes can most easily be accomplished by using target-specific immobilized antibody or PTM specific affinity binding. We offer several kits targeting phosphorylated proteins, alpha- linked mannose and terminal glucose residues, N-acetyl glucosamine, polyubiquinated proteins, ATP binding sites, and active serine hydrolase enzymes. Choose the right protein enrichment kit for your application using the following link (http://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-purification/protein-enrichment.html).

For Research Use Only. Not for use in diagnostic procedures.