missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSAT (tris-(succinimidyl)aminotriacetate)

Catalog No. pi33063
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI33063 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI33063 Supplier Thermo Scientific™ Supplier No. LSG33063

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A 3-way, short-arm (4.2A) amine-to-amine crosslinker; tris (succinimidyl) aminotriacetate, composed of three NHS-ester groups on short spacer arms; membrane permeable.

Thermo Scientific™ Pierce TSAT is tris(succinimidyl)aminotriacetate, a novel trifunctional crosslinker that contains three short spacers arms linked to a central nitrogen atom, each ending in an amine-reactive NHS-ester group.

Highlights:

  • Reactive group: maleimide
  • Reactive toward: sulfhydryl groups

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description TSAT
Molecular Weight (g/mol) 482.36
PEGylated No
Spacer Arm Length 4.2 Å
Content And Storage Upon receipt store at 4°C protected from moisture.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homotrifunctional
Reactive Moiety NHS Ester
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for TSAT (tris-(succinimidyl)aminotriacetate)?

TSAT (tris-(succinimidyl)aminotriacetate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.