missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tris

Catalog No. PI17926
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI17926 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI17926 Supplier Thermo Scientific™ Supplier No. LSG17926

Perform a variety of protein and molecular biology methods. Thermo Scientific™ Pierce™ Tris is a standard Tris buffering reagent available in a 1kg pack.

Thermo Scientific™ Pierce Tris is a standard reagent for preparation of many molecular biology buffers.

Additional Information and Online Ordering

Thermo Scientific™ Pierce™ Tris is a standard reagent for preparation of many molecular biology buffers.

  • ≥99.8% pure
  • White crystalline powder
  • Optimal buffering between pH 7 and 9

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Product Type Tris
Purity ≥99.8%
Quantity 1 kg
Form Powder

For Research Use Only. Not for use in diagnostic procedures.