missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution

Catalog No. PI88325
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88325 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88325 Supplier Thermo Scientific™ Supplier No. 88325

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calibrate Triple Stage Quadrupole (TSQ) mass spectrometers using a mixture of high purity, ionizable components specifically designed for positive mode calibration.

Thermo Scientific™ Pierce Triple Quadrupole Calibration Solution is a mixture of high purity, ionizable components specifically designed for positive mode calibration of Thermo Scientific™ Triple Stage Quadrupole instruments.

The Pierce Triple Quadrupole Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the required routine calibration to maintain the robust performance of Thermo Scientific Mass Spectrometers. The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

Highlights

  • Strong peaks — mixture of three tyrosine isotopes in methanol/formic acid solution
  • Ready-to-use — load the reference standard into a syringe and inject into the instrument
  • High-purity — mass spectrometry grade reagents in a a non-leachable container
  • Stable — store at 2° to 8°C for more than one year

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Calibration Solution
Product Line Pierce
Content And Storage Store between 2°C and 8°C.
Starting Material Small Molecules
Final Product Type Small Molecules
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 10 mL
Form Liquid
Which instruments can be calibrated using the Pierce Triple Quadrupole Calibration Solution (Cat. No. 88325)?

The Pierce Triple Quadrupole Calibration Solution (Cat. No. 88325) is a mixture of high purity, ionizable components specifically designed for positive mode calibration of Thermo Scientific Triple Stage Quadrupole instruments such as TSQ Quantum, TSQ Discovery, TSQ Quantum Ultra, TSQ Quantum Access, TSQ Vantage, TSQ Endura, and TSQ Quantiva Series Mass Spectrometer instruments.

For Research Use Only. Not for use in diagnostic procedures.