missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Trypsin, TPCK Treated

Catalog No. PI20233
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20233 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20233 Supplier Thermo Scientific™ Supplier No. 20233

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.

Thermo Scientific™ Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.

Thermo Scientific Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.

Features of Thermo Scientific Pierce TPCK Trypsin
• Cleaves at carboxyl side of arginine and lysine residues
• Digestion conditions: pH 7.5–9.0, 37°C

Trypsin has a wide range of applications including amino acid analysis and protein sequencing, mapping and structural studies. Enzymes such as trypsin and chymotrypsin have become important tools in sequencing studies since they are highly selective in their cleavage of peptide bonds. Trypsin cleaves only those peptide bonds in which the carboxyl group is contributed by a lysine or an arginine residue, regardless of the length or amino acid sequence of the chain. Our Immobilized TPCK Trypsin can be substituted for free trypsin in many application and is advantageous because it minimizes autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the trypsin. Immobilized trypsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity.

Applications:
• Removal of adherent cells from tissue culture flasks
• Preparing tryptic fragments for Edman degradation sequencing
• Immobilized trypsin can be used to purify soybean trypsin inhibitor
• Use our Pierce Trypsin, MS Grade (Cat No. 90057) for mass spectrometry workflows

Trypsin is a 23.8kDa pancreatic serine endoprotease derived from trypsinogen, an inactive precursor zymogen, after enzymatic removal of an n-terminal leader sequence by enterokinase. Once some trypsin has been formed it can catalyze the conversion of more trypsinogen into its catalytically-active form. Trypsin is treated with L-1-tosylamido-2-phenylethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotrypsin activity without affecting trypsin activity.

Dissolving 0.25 g of trypsin in 100 mL of chilled HBSS for a 0.25% w/v solution is recommended. Mix solution until homogenous and pH is 7.4 to 7.8. Filter the solution through a low protein-binding filter such as the Nalgene™ Rapid-Flow™ Sterile Disposable Filter Units with PES Membranes (Cat No. 567-0010) and store -5°C to -20°C.

Related Products
Immobilized Trypsin, TPCK Treated (Agarose Resin) (Cat. No. 20230)

Specifications

Description Pierce TPCK Trypsin
Content And Storage Store in original container protected from direct sunlight in a dry, cool, and well-ventilated area from 2–8°C.
Form Powder
Quantity 50 mg
Product Type Trypsin
Do you have a protocol for making stock solution of Trypsin, TPCK Treated (Cat. No. 20233)?

This product should primarily be used as a powder directly in the digestion buffer.
Materials:
Digestion Buffer
0.1 M Ammonium carbonate (NH4)HCO3, pH 8.0.

Procedure:
1. Dissolve approximately 10 mg of the protein (=substrate) in 1 ml of Digestion Buffer.
2. Add sufficient TPCK Trypsin to the protein solution to have an enzyme: substrate ratio between 1:20 and 1:100 (w/w).
3. Incubate sample for 2–18 hours at 37 degrees C.
Note: The exact incubation time and enzyme: substrate ratio will have to be determined for each protein individually.

References:
Walsh, K.A., Neurath, H. (1964). Trypsinogen and chymotrypsinogen as homologous proteins. Proc. Natl. Acad. Sci. USA, 52, p. 884.
Cunningham, L.W. (1954). Molecular-kinetic properties of crystalline diisopropyl phosphoryl trypsin. J. Biol. Chem. 211, p.13.
Kostka, V., Carpenter, F. H. (1964). Inhibition of chymotrypsin activity in crystalline trypsin preparations. J. Biol. Chem. 239, 1799–1803.

For Research Use Only. Not for use in diagnostic procedures.