missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immobilized Trypsin, TPCK Treated (Agarose Resin)

Catalog No. PI20230
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20230 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20230 Supplier Thermo Scientific™ Supplier No. LSG20230
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleave the carboxyl-side of Arg and Lys residues with trypsin that isTPCK-treated to block chymotrypsin activity; its immobilized form allows enzyme removal after digestion.

Thermo Scientific™ Pierce TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.

Highlights:

  • Immobilized TPCK Trypsin eliminates trypsin contamination in digests
  • TPCK inhibits chymotrypsin activity

Specifications

Description Pierce Immobilized Trypsin, TPCK Treated
Content And Storage Contents: Immobilized TPCK Trypsin, 2 mL of settled gel supplied as a 50% slurry containing glycerol and 0.05% sodium azide.
Storage: Upon receipt store at 4°C.
Product is shipped at ambient temperature.
Form Liquid Suspension
Quantity 2 mL
Product Type Trypsin

For Research Use Only. Not for use in diagnostic procedures.