missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ TMB ELISA Substrate Solutions

Catalog No. PI34029
Change view
Click to view available options
Quantity:
400 mL Kit
250 mL
1 L
Substrate:
Slow TMB-ELISA Substrate
TMB Substrate
Turbo TMB-ELISA Substrate
Ultra TMB-ELISA Substrate
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Substrate
PI34029 1 L Ultra TMB-ELISA Substrate
PI34024 250 mL Slow TMB-ELISA Substrate
PI34022 250 mL Turbo TMB-ELISA Substrate
PI34028 250 mL Ultra TMB-ELISA Substrate
PI34021 400 mL Kit TMB Substrate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PI34029 Supplier Thermo Scientific™ Supplier No. 34029

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce TMB ELISA Substrates detect horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.

TMB (3,3',5,5' tetramethylbenzidine) is a chromagen that yields a blue color when oxidized with hydrogen peroxide (catalyzed by HRP) with major absorbances at 370 nm and 652 nm. The color then changes to yellow (measured at 450 nm) upon addition of sulfuric or phosphoric acid to stop the reaction. TMB is very sensitive and more quickly oxidized than other HRP substrates, resulting in faster color development. Unlike other commercially available substrates, these products contain no DMF or DMSO.

There are three formulations that differ primarily in their sensitivities. Slow TMB is intermediate in sensitivity, ideal for kinetic readings. The sensitivity of the Turbo TMB compares to that of OPD used at approximately 1 mg/mL. Ultra TMB-ELISA produces the highest signal-to-noise ratio and sensitivity in the picogram range.

Thermo Scientific Pierce 1-Step TMB ELISA Substrates detects horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.

Features of 1-Step TMB ELISA Substrates:
• Available in traditional or ready-to-use formulations
• No additional reagents or filtering required
• Non carcinogenic
• Various levels of sensitivity and development speed to suit any assay
• No DMF or DMSO present in the reagents

TMB is the most popular chromogenic substrate for HRP detection in ELISA and is available in several formats. The Pierce 1-Step TMB Substrates are one-component substrates that require no preparation before use. Unlike other commercially available substrates, these products contain no DMF or DMSO.

Thermo Scientific Pierce 1-Step Ultra TMB yields the greatest sensitivity among the TMB substrates, followed by 1-Step Turbo TMB and 1-Step Slow TMB. The sensitivity of the 1-Step Turbo TMB is similar to OPD-based substrates formulated at approximately 1 mg/mL. 1-Step Slow TMB has intermediate sensitivity—more sensitive than ABTS, but less sensitive than o-phenylenediamine dihydrochloride (OPD) or 1-Step Turbo TMB. The 1-Step Slow TMB is an ideal substrate for kinetic studies.

TMB (3,3',5,5'-tetramethylbenzidine) is a chromogen that yields a blue color when oxidized, typically as a result of oxygen radicals produced by the hydrolysis of hydrogen peroxide by HRP. For kinetic or non-stopped ELISA assays, the TMB chromogen has maximal absorbances at 370 nm and 652 nm. The color then changes to yellow with the addition of sulfuric or phosphoric acid with maximum absorbance at 450 nm. A green reaction product may result from partial conversion to the yellow product from the blue intermediate. TMB is very sensitive ELISA substrate and is more quickly oxidized than other HRP substrates, resulting in faster color development.

Specifications

Product Type ELISA Substrate
Quantity 1 L
Detection Method Colorimetric
Product Line 1-Step
Substrate Ultra TMB-ELISA Substrate
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
Content And Storage Upon receipt store at 4°C, protected from moisture.
Shipping Condition Ambient
Are all your TMB substrates transparent and do they all turn blue after reacting with the HRP enzyme of an antibody?

No, we offer different TMB substrates for different applications.
We recommend the following TMB substrates for this particular application:
- eBioscience TMB Solution (1X) (Cat. No. 00-4201-56)
- (For higher sensitivity) 1-Step Ultra TMB-ELISA Substrate Solution (Cat. No. 34029)

What is the shelf-life of 1-Step Ultra TMB-ELISA Substrate Solution?

The 1-Step Ultra TMB-ELISA Substrate Solution is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

My lab doesn't have a fluorescent or chemiluminescent plate reader and I need the most sensitive substrate for Horseradish Peroxidase (HRP). Which of your colorimetric substrates should I use?

The 1-Step Ultra TMB-ELISA is our most sensitive substrate for HRP in plate-based assays. It can detect down to ~ 2 pg/well or 20 pg/mL.

Can the 1-Step Ultra TMB-ELISA Substrate Solution be used as a precipitating chromogen for Western Blot, Immunohistochemistry and immunocytochemistry?

No. This product is formulated to generate a soluble, colored product that absorbs light at 450 nm. When used properly, there will be no precipitate, therefore, this substrate is only appropriate for plate-based assays, such as ELISA.

Is the 1-Step Ultra TMB-ELISA Substrate Solution an appropriate substrate for kinetic assays?

No. Although the non-stopped substrate will turn a blue color in the presence of Horseradish Peroxidase and absorb light at 307 nm and 652 nm, color development proceeds very quickly. In the time that it takes to read the full plate, the absorbance of the first wells will have changed. It is better to stop the the color development in all wells at approximately the same time point with an acidic soluton, turning it yellow and then read the absorbance at 450 nm.