missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ TCEP-HCl, No-Weigh™ Format

Catalog No. pi20491
Change view
Click to view available options
Quantity:
10 x 1 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20491 10 g
PI20490 1 g
PIA35349 10 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI20491 Supplier Thermo Scientific™ Supplier No. 20491
Only null left

Reduce protein and peptide disulfide bonds using this thiol-free, pure, crystalline tris (2-carboxyethyl) phosphine hydrochloride, TCEP (CAS 5961-85-3).

Thermo Scientific™ Pierce TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable.

Highlights:

  • Reduces even the most stable water-soluble alkyl disulfides
  • Effective at room temperature and pH 5 in less than five minutes
  • Does not require removal before reaction with maleimide or haloacetyl compounds
  • Water solubility of 310g/L
  • Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups in proteins

Specifications

Content And Storage Upon receipt store at room temperature in sealed container to prevent oxidation. Product shipped at ambient temperature.
Product Line Pierce
Form Solid
Quantity 10 g
Solubility Water
Product Type Disulfide Reducing Reagent

For Research Use Only. Not for use in diagnostic procedures.