missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format

Catalog No. pia39268
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 x 2 mg
50 mg
Catalog No. Quantity
PIA39268 10 x 2 mg
PI22322 50 mg
PI22122 1 g
3 options

Catalog No. PIA39268

Supplier: Thermo Scientific™ A39268

Only null left

Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker made of NHS-ester and maleimide groups at ends of a cyclohexane-containing, 8-atom spacer arm.

Thermo Scientific™ Pierce Sulfo-SMCC is a very popular, water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 Ås).

  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Water-soluble (compare to SMCC), so crosslinking can be done in physiologic solutions
  • High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
  • Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description Sulfo-SMCC
Molecular Weight (g/mol) 436.37
PEGylated No
Spacer Arm Length 8.3 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line No-Weigh
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short (<10 Å)
Form Solid
Quantity 10 x 2 mg
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.