missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Sulfo-SANPAH, No-Weigh™ Format
Description
Thermo Scientific™ Pierce Sulfo-SANPAH is a long-arm (18.2 Å) crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.
Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.
- Reactive groups: sulfo-NHS ester and nitrophenyl azide
- Reactive towards: amino groups and any nucleophile
- Non-cleavable
- N-Sulfosuccinimidyl-6-(4'-azido-2'-nitrophenylamino) hexanoate
- Optimal photolysis occurs at 320-350nm; minimizes damage to biomolecules by irradiation
- Water soluble; non-cleavable
Specifications
Specifications
| Chemical Reactivity | Amine-Nonselective |
| Cleavable | By Thiols |
| Molecular Weight (g/mol) | 492.4 |
| PEGylated | No |
| Spacer Arm Length | 18.2 Å |
| Content And Storage | Upon receipt store at -20°C protected from light and moisture. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | No-Weigh, Pierce |
| Labeling Method | Chemical Labeling |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.