missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Sulfo-SANPAH, No-Weigh™ Format

Catalog No. PIA35395
Change view
Click to view available options
Quantity:
50 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA35395 10 x 1 mg
PI22589 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA35395 Supplier Thermo Scientific™ Supplier No. A35395
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce Sulfo-SANPAH is a long-arm (18.2 Å) crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.

Thermo Scientific™ Pierce Sulfo-SANPAH is a long-arm (18.2 Å) crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Reactive groups: sulfo-NHS ester and nitrophenyl azide
  • Reactive towards: amino groups and any nucleophile
  • Non-cleavable
  • N-Sulfosuccinimidyl-6-(4'-azido-2'-nitrophenylamino) hexanoate
  • Optimal photolysis occurs at 320-350nm; minimizes damage to biomolecules by irradiation
  • Water soluble; non-cleavable

Specifications

Chemical Reactivity Amine-Nonselective
Cleavable By Thiols
Molecular Weight (g/mol) 492.4
PEGylated No
Spacer Arm Length 18.2 Å
Content And Storage Upon receipt store at -20°C protected from light and moisture.
Cell Permeability No
Shipping Condition Ambient
Product Line No-Weigh, Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Sulfo-NHS Ester, Nitrophenyl Azide
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 10 x 1 mg
Solubility Water
Format Standard, Single-use
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for Sulfo-SANPAH?

Sulfo-SANPAH is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

What are the reactive groups on Sulfo-SANPAH and which groups do they react with?

Sulfo-SANPAH is a heterobifunctional crosslinker that has a sulfo-NHS ester and a photoactivatable nitrophenyl azide group. The sulfo-NHS ester reacts with primary amines in pH 7-9 buffers to form stable amide bonds. When exposed to UV light, nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).

For Research Use Only. Not for use in diagnostic procedures.