missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-LC-SPDP (sulfosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)

Catalog No. PI21650
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21650 Supplier Thermo Scientific™ Supplier No. 21650

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink amines to sulfhydryls with this water-soluble sulfo-NHS-ester and pyridyldithiol groups on 10-atom spacer arms to form cleavable disulfide bonds with cysteines.

Thermo Scientific™ Pierce Sulfo-LC-SPDP is a water-soluble, long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteines.

Highlights:

  • Forms cleavable disulfide bonds with sulfhydryls
  • Releases a detectable byproduct after reacting with free sulfhydryl groups; by measuring the release of pyridine-2-thione at 343nm, the reaction progression can be monitored
  • Reactive groups: sulfo-NHS ester and pyridyldisulfide
  • Reactive toward: amino and sulfhydryl groups

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description Sulfo-LC-SPDP
Molecular Weight (g/mol) 527.57
PEGylated No
Spacer Arm Length 15.7 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Pyridyldisulfide, Sulfo-NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for Sulfo-LC-SPDP (sulfosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)?

Sulfo-LC-SPDP (sulfosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.