missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-EGS (ethylene glycol bis(sulfosuccinimidyl succinate))

Catalog No. pi21566
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21566 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21566 Supplier Thermo Scientific™ Supplier No. LSG21566

Crosslink amines to amines with a crosslinker composed of sulfo-NHS-ester groups and hydroxylamine-cleavable spacer arm; water-soluble for cell-surface protein crosslinking.

Thermo Scientific™ Pierce Sulfo-EGS is a water-soluble crosslinker that contains amine-reactive sulfo-NHS-ester ends around a 12-atom spacer arm, which can be cleaved by treatment with hydroxylamine at pH 8.5.

Highlights:

  • Water-soluble
  • Cleavable at pH 8.5 using hydroxylamine and heat
  • Lactose dehydrogenase retained 60% of its activity after reversible crosslinking
  • Reactive groups: sulfo-NHS esters (homobifunctional)
  • Reactive toward: amino groups

Specifications

Chemical Reactivity Amine-Amine
Cleavable By Hydroxylamine
Description Sulfo-EGS
Molecular Weight (g/mol) 660.45
PEGylated No
Spacer Arm Length 16.1 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Sulfo-NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for Sulfo-EGS (ethylene glycol bis(sulfosuccinimidyl succinate))?

Sulfo-EGS (ethylene glycol bis(sulfosuccinimidyl succinate)) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.