Learn More
Thermo Scientific™ Pierce™ Streptavidin Agarose
Description
Thermo Scientific™ Pierce Streptavidin Resins comprise a wide selection of high-quality affinity beads and columns for nearly any kind of biotin-based purification procedure.
Additional Information and Online Ordering
Highlights:
Proven coupling chemistries – the methods used to prepare these resins are superior to typical cyanogen bromide (CNBr) immobilization, providing high functionality, low nonspecific binding and excellent leach-resistance
Versatile resins – Agarose (polysaccharide-based) and UltraLink™ Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods
Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)
Superior performance – High Capacity Resin has 2 to 3 times higher binding capacity than alternative supplier products, allowing use of small amounts of resin for experiments
Item-Specific Features:
- Agarose Resins are crosslinked 6% beaded agarose
- UltraLink Resins beaded acrylamide support (UltraLink Biosupport)
- Plus and High Capacity Resins have higher loading (i.e. density) of immobilized streptavidin protein than the regular forms
Recommended for:
Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies; Cell-surface labeling with biotinylation reagents, followed by precipitation with immobilized streptavidin; Purification of cell-surface glycoproteins using biotinylated Concanavalin A; Recovery of single-stranded DNA for dideoxy sequencing
Specifications
Specifications
| Content And Storage | Upon receipt store at 4°C. Do not freeze the resin. Product is shipped at ambient temperature. |
| Particle Size | 45 to 165 μm |
| Product Line | Pierce |
| Quantity | 10 mL |
| Protein Form | Recombinant |
| Target | Biotin-Tagged Molecule |
| Type | Agarose |
| Stationary Phase | Streptavidin |
| Sufficient For | Binding 15 to 28 μg Biotin/mL Resin (1 to 3 mg Biotinylated BSA/mL Resin) |
| Format | Bottle |
| Show More |
Frequently Asked Questions (FAQs)
The average particle size for the Pierce Streptavidin Agarose beads is 45 to 165 µm.
The antigen:antibody ratio will need to be determined empirically since one cannot be sure of the amount of specific antibody against the antigen present in the plasma. The starting point is the binding capacity of the High Capacity Streptavidin Agarose Resin (Cat. No. 20347) which is 1-3 mg of biotinylated BSA/mL of settled resin. The binding capacity will vary depending on the molecular weight of the biotinylated antigen. Let's assume the molecular weight of the antigen is close to that of BSA (66.4 kDa), you can use an average of 2 mg antibody or antigen/mL of resin to do the calculation. Determine how much resin they want to use and calculate how much biotinylated protein is needed and then empirically determine how much plasma is needed.
We guarantee the functionality of the product for one year from the date of shipment, if stored as recommended.
Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.
For Research Use Only. Not for use in diagnostic procedures.