missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Stop Solution for TMB Substrates

Catalog No. PIN600
Change view
Click to view available options
Quantity:
55 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIN600 55 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIN600 Supplier Thermo Scientific™ Supplier No. N600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stop TMB-based ELISA detection with this 0.16M sulfuric acid for use with the ELISA substrate 3,3',5,5' - tetramethylbenzidine (TMB).

Thermo Scientific™ Pierce Stop Solution for TMB is 0.16M sulfuric acid for use with the ELISA substrate 3,3',5,5' - tetramethylbenzidine (TMB).

TMB Stop Solution is 0.16M sulfuric acid for use with the ELISA substrate 3,3«,5,5«-tetramethylbenzidine (TMB). Addition of sulfuric acid stop solution changes the color from blue to yellow, stabilizing the color development to enable accurate measurement of the intensity at 450nm using a spectrophotometer or plate reader.
  • Size: 55mL

  • Sufficient For: five 96-well microplates

  • Application: ELISA

Instructions: Following TMB incubation, add an amount of stop solution equal to the amount of TMB substrate added to each well of the ELISA plate (typically 50 to 100μL per well). Use at full strength.

Specifications

Product Type Stop Solution
Quantity 55 mL
Form Liquid
For Use With (Application) ELISA
Content And Storage Store at 2–8°C.

For Research Use Only. Not for use in diagnostic procedures.