missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SPDP (succinimidyl 3-(2-pyridyldithio)propionate)

Catalog No. pi21857
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21857 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21857 Supplier Thermo Scientific™ Supplier No. 21857

Crosslink amines to sulfhydryls with this crosslinker composed of NHS-ester and pyridyldithiol groups on 3-atom spacer arm; forms cleavable disulfide bonds with cysteines.

Thermo Scientific™ Pierce SPDP is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls.

Highlights:

  • Widely used in immunochemistry; conjugates used in drug carrier systems, antibody production and enzyme immunoassays
  • Can be used as a protein thiolation reagent
  • Reactive groups: NHS ester and pyridyldisulfide
  • Reactive toward: amino and sulfhydryl groups

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description SPDP
Molecular Weight (g/mol) 312.36
PEGylated No
Spacer Arm Length 6.8 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Pyridyldisulfide
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard, Premium-grade
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for SPDP (succinimidyl 3-(2-pyridyldithio)propionate)?

SPDP (succinimidyl 3-(2-pyridyldithio)propionate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.