missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl

Catalog No. PI28365
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28365 1 kg
PI28364 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI28365 Supplier Thermo Scientific™ Supplier No. 28365

Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.

Thermo Scientific Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).

Features of Sodium Dodecyl Sulfate (Lauryl):

  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
  • Common component of cell lysis buffers

This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).

Properties of SDS (values for pure C12):

  • Molecular Weight: 288.5 g
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 62
  • Micelle Molecular Weight: 18,000 g
  • Critical Micelle Concentration (CMC): 6 to 8 mM (0.1728 to 0.2304%, w/v)
  • Cloud Point: >100°C
  • Dialyzable: No

Specifications for Sodium Dodecyl Sulfate:

  • Visual: White powder, free of foreign material.
  • Solubility: 10% (aq, w/v) solution must be clear, colorless to slightly yellow.
  • Chain length: C12 >60%; C14 = 20 to 35%; C16 <10%; C10 and C18 <1% each

Related Products

  • Sodium Dodecyl Sulfate (SDS), C12 (Cat. No. 28312)
  • Sodium Dodecyl Sulfate (SDS), 20% Solution (Cat. No. 62202)

Specifications

Color White
Product Type Detergent
Solubility 10% (aq, w/v) solution must be clear, colorless to slightly yellow
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Powder
Reagent Type Detergent (Pure)
Quantity 1 kg
Why is LDS (lithium dodecyl sulfate) used in the 4X NuPAGE sample buffer instead of SDS?

SDS in a 4X sample buffer concentrate tends to precipitate from solution and to make the solution viscous and difficult to pipette. The LDS is much more soluble.

Are detergents denaturing or non-denaturing with respect to protein structure?

Ionic detergents, or those that carry a charge, are the most likely to be denaturing to proteins. Denaturing detergents can be anionic such as sodium dodecyl sulfate (SDS) or cationic such as ethyl trimethyl ammonium bromide. These detergents totally disrupt membranes and denature proteins by breaking protein-protein interactions through changes in the three-dimensional structure of the proteins. Nondenaturing detergents can be divided into nonionic detergents (i.e., Triton X-100), bile salts (i.e., cholate), and zwitterionic detergents (i.e., CHAPS).

What are detergents?

Detergents are amphipathic molecules containing both a nonpolar “tail” having aliphatic or aromatic character, and a polar “head”. The ionic character of the polar head group forms the basis for broad classification of detergents as ionic, nonionic, or zwitterionic.

How does detergent-based cell lysis work?

Detergents are amphipathic molecules, meaning they contain both a nonpolar “tail” having aliphatic or aromatic character and a polar “head”. Like the components of biological membranes, detergents have hydrophobic-associating properties as a result of their nonpolar tail groups. Nevertheless, detergents are themselves water soluble.

Consequently, detergent molecules allow the dispersion (miscibility) of water-insoluble, hydrophobic compounds into aqueous media, including the extraction and solubilization of membrane proteins. Detergent monomers solubilize membrane proteins by partitioning into the membrane bilayer. With increasing amounts of detergents, membranes undergo various stages of solubilization.

What types of detergents are available for cell lysis?

Detergents can be denaturing or non-denaturing with respect to protein structure. Denaturing detergents can be anionic such as sodium dodecyl sulfate (SDS) or cationic such as ethyl trimethyl ammonium bromide. These detergents totally disrupt membranes and denature proteins by breaking proteinprotein interaction. These detergents are considered harsh. Non-denaturing detergents can be divided into nonionic detergents (i.e., Triton X-100), bile salts (i.e., cholate), and zwitterionic detergents (i.e., CHAPS). These detergents do not denature proteins and do not break protein-protein interactions. These detergents are considered mild.

Why does the method of cell lysis matter?

Cell lysis is the first step in cell fractionation, organelle isolation, and protein extraction and purification. As such, cell lysis opens the door to a myriad of proteomics research methods. Many techniques have been developed and used to obtain the best possible yield and purity for different species of organisms, sample types (cells or tissue), and target molecule or subcellular structure. Subcellular fractionation and protein enrichment are important methods in the rapidly growing field of proteomics. Isolation of subcellular fractions and concentration of proteins in low abundance allow for more efficient identification and study of proteins of interest. Examples are the isolation of integral membrane proteins and nuclear proteins.

What methods of cell lysis are available?

Historically, physical lysis was the method of choice for cell disruption and extraction of cellular contents; however, it often requires expensive, cumbersome equipment and involves protocols that can be difficult to repeat due to variability in the apparatus (such as loose-fitting compared with tight-fitting homogenization pestles). Also, traditional physical disruption methods are not conducive for high-throughput and smaller volumes typical of modern laboratory research.
In recent years, detergent-based cell lysis methods have become the norm. Through empirical testing by trial and error, different detergent-based solutions composed of particular types and concentrations of detergents, buffers, salts and reducing agents have been developed to provide the best possible results for particular species and types of cells. Detergents have both lysing and solubilizing effects.


For Research Use Only. Not for use in diagnostic procedures.