missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene)

Catalog No. PI21558
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21558 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21558 Supplier Thermo Scientific™ Supplier No. LSG21558

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink amines to sulfhydryls with this crosslinker containing NHS-ester and pyridyldithiol groups at the ends of an aromatic, 6-atom (11.2A) spacer arm.

Thermo Scientific™ Pierce SMPT is a long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form strong (hindered) yet cleavable (reducible) disulfide bonds with cysteine sulfhydryls.

Highlights:

  • Contains a hindered disulfide bond; forms stable immunotoxins
  • In vitro, an SMPT conjugate was as effective as conjugates formed with SPDP and 2-iminothiolane
  • Reactive groups: NHS ester and pyridyldisulfide
  • Reactive toward: amino and sulfhydryl groups

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description SMPT
Molecular Weight (g/mol) 388.46
PEGylated No
Spacer Arm Length 11.2 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Pyridyldisulfide
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene)?

SMPT (4-succinimidyloxycarbonyl-alpha-methyl-α(2-pyridyldithio)toluene) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.