missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SMCC

Catalog No. PIA35394
Change view
Click to view available options
Quantity:
50 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA35394 10 x 1 mg
PI22360 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIA35394 Supplier Thermo Scientific™ Supplier No. A35394
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No-Weigh™ Format

Thermo Scientific™ Pierce SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm (8.3 Ås).

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Reactive groups: NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO); compare to Sulfo-SMCC
  • High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
  • Cyclohexane bridge confers added stability to the maleimide group making SMCC an ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C
Applications:
  • Enzyme labelling of antibodies - both enzyme and antibody specificity can be preserved
  • Create specific bioconjugates via one- or two-step crosslinking reactions
  • Create sulfhydryl-reactive, maleimide-activated carrier proteins for coupling haptens

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Molecular Weight (g/mol) 334.32
PEGylated No
Spacer Arm Length 8.3 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line No-Weigh, Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 10 x 1 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)?

SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

What is the difference between SMCC and Sulfo-SMCC?

SMCC is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane-stabilized spacer arm (8.3 angstroms). Sulfo-SMCC is a similar compound but it has a sulfonate group on the NHS ring which makes it water-soluble whereas SMCC is water-insoluble and needs to be dissolved first in an organic solvent like DMF or DMSO. Subsequent dilution into aqueous reaction buffer is generally possible, and most protein reactants will remain soluble if the final concentration of organic solvent is less than 10%.

For Research Use Only. Not for use in diagnostic procedures.