missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SDS-Out™ SDS Precipitation Kit

Catalog No. PI20308
Change view
Click to view available options
Quantity:
10 mL kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI20308 10 mL kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI20308 Supplier Thermo Scientific™ Supplier No. LSG20308

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quickly remove SDS from small-volume (0.1 to 10mL) protein samples with this SDS precipitation kit.

Perform convenient removal of SDS from small-volume (0.1 to 10mL) protein samples with Thermo Scientific™ SDS-Out SDS Precipitation Reagent.

The simple reagent and kit effectively remove excess SDS from small protein samples. Sodium dodecyl sulfate (SDS) is a detergent typically used in cell lysis reagents and other protein solubilization buffers. Traditional methods for SDS removal include prolonged dialysis, anion exchange chromatography, affinity columns or and acetone precipitation. By comparison, the SDS-Out Precipitation Reagent is relatively easy to use and provides excellent protein recovery.

Highlights:

  • Convenient removal of SDS from microliter-volume samples
  • Use for samples containing 0.1 to 1% SDS
  • Minimal dilution of small-volume samples
  • Protein recovery is 90 to 100% (based on tests with BSA, cytochrome C, soybean trypsin inhibitor, ovalbumin, ribonuclease A, myoglobin and human IgG)

Includes:

Spin cup and collection tube accessories for use with the SDS-Out Reagent

Specifications

Product Type SDS Precipitation Kit
Content And Storage Sufficient For: Precipitation of SDS from 200 mL of protein solution
• SDS Precipitation Reagent, 10 mL
• Spin Cup Columns, 12 columns
• Collection Tubes, 12 tubes

Upon receipt, store components at 4°C.
Column Type Precipitation, Filtration, n/a
Product Line SDS-Out
Purification Target Detergent Removal
Sufficient For 200 mL Protein Solution
Quantity 10 mL kit
Format Kit, Microcentrifuge, Spin Column

For Research Use Only. Not for use in diagnostic procedures.