missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SATA (N-succinimidyl S-acetylthioacetate)

Catalog No. PI26102
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26102 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26102 Supplier Thermo Scientific™ Supplier No. 26102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Short (2.8A), amine-reactive (NHS ester) sulfhydryl-addition reagent; creates stable (protected yet exposable) -SH group for protein crosslinking strategies.

Thermo Scientific™ Pierce SATA is a short-chain (2.8 Å spacer arm) reagent for covalent modification of primary amines and addition of a stable yet exposable sulfhydryl group to facilitate heterobifunctional crosslinking strategies.

Highlights:

  • Reacts with primary amines to add protected sulfhydryls
  • An easy deprotection step generates a free sulfhydryl group

Specifications

Chemical Reactivity Amine
Content And Storage Upon receipt store desiccated at -20°C. Product is shipped at ambient temperature.
Label or Dye Sulfhydryl Group
Label Type Conjugating Groups
Product Line Pierce
Crosslinker Type Heterobifunctional
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Short (<10 Å)
Form Powder
Reagent Type SATA
Quantity 50 mg
Solubility DMF, DMSO
Product Type Crosslinker
Show More Show Less
What is the best way to remove unreacted SATA (Cat. No. 26102) after peptide modification?

The best way for removing unreacted SATA (Cat. No 26102) would be HPLC. Another possibility would be to use desalting columns. In case the peptide is bigger than 1.8 kDa, you would like to use the Pierce Polyacrylamide Desalting Columns, 1.8K MWCO, 5 mL (Cat. No 43426).

For Research Use Only. Not for use in diagnostic procedures.