missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SAT(PEG)4 (PEGylated N-succinimidyl S-acetylthioacetate)

Catalog No. PI26099
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26099 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26099 Supplier Thermo Scientific™ Supplier No. 26099

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Long (18.2A), pegylated form of SATA, an amine-reactive (NHS ester) sulfhydryl-addition reagent; contains 4-unit polyethylene glycol spacer arm.

Thermo Scientific™ Pierce SAT(PEG)4 is a long-arm (18.2 Ås), pegylated form of SATA, a sulfhydryl-addition reagent for covalent modification of primary amines to facilitate controlled, heterobifunctional crosslinking.

SAT(PEG)4 extends the flexibility, solubility and reach of other S-acetylthio (SAT) reagents for sulfhydryl modification of amine-containing molecules. SAT(PEG)4 has the same form as SATA and SATP, which are widely used as part of protein conjugation strategies. Two purified proteins (e.g., antibody and enzyme) are conjugated after modifying one protein with an SAT reagent and the other protein with a maleimide-activating crosslinker such as Sulfo-SMCC (Part No. 22322). Because SAT reagents add a protected sulfhydryl, the modified protein can be stored indefinitely before treating it to expose the sulfhydryl and performing the final sulfhydryl-directed conjugation reaction.The four-unit polyethylene glycol (PEG) group in SAT(PEG)4 provides increased hydrophilicity and spacer-arm length relative to SATA and SATP. When used in combination with a long-arm PEG-containing crosslinker like SM(PEG)12 (Part No. 22112), SAT(PEG)4 can be used to crosslink proteins with long spacers.

Highlights:

  • Reacts with primary amines to add protected sulfhydryls
  • An easy deprotection step generates a free sulfhydryl group
  • Water-soluble polyethylene glycol (PEG) spacer arm enhances solubility of labeled protein

Specifications

Chemical Reactivity Amine
Content And Storage Upon receipt store desiccated at -20°C. Product shipped at ambient temperature.
Label or Dye Sulfhydryl Group
Label Type Conjugating Groups
Product Line Pierce
Crosslinker Type Heterobifunctional
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Long, PEGylated
Form Liquid
Quantity 100 mg
Solubility DMF, DMSO
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for SAT(PEG)4 (PEGylated N-succinimidyl S-acetylthioacetate)?

SAT(PEG)4 (PEGylated N-succinimidyl S-acetylthioacetate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.