missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Reserpine Standard for LC-MS

Catalog No. PI88326
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88326 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88326 Supplier Thermo Scientific™ Supplier No. LSG88326

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An exact solution of reserpine specifically designed for performance evaluation of mass spectrometers, including the Thermo Scientific™ ion trap and TSQ series of instruments.

Thermo Scientific™ Pierce Reserpine Standard for LC-MS is an exact solution of reserpine specifically designed for performance evaluation of mass spectrometers, including the Thermo Scientific ion trap and TSQ series of instruments.

Additional Information and Online Ordering

The Pierce Reserpine Standard for LC-MS is a pre-diluted liquid formulation ideal to quickly perform installation specifications of Thermo Scientific (and other) mass spectrometers. The standard is provided in a concentration of 100pg/μL in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5 x 1mL glass amber vials with PTFE-lined screw caps. The Pierce Reserpine Standard for LC-MS is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications.

Highlights:

  • Convenient – provided pre-diluted and requires minimal additional dilutions to reach target concentration for injection

  • Safe handling – unlike glass ampules, cap can be easily removed and replaced when withdrawing solution

  • High-purity – mass spectrometry grade reagent in non-leachable screw-cap vials

  • Stable – store at 4°C for up to one year

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Reserpine Standard
Product Line Pierce
Content And Storage Store in a cool, dry, well-ventilated area, protected from direct sunlight.
Starting Material Peptides, Small Molecules
Final Product Type Peptides, Small Molecules
Detection Method Mass Spectrometry
Workflow Step Calibration
Quantity 5 x 1 mL
Form Liquid
What is the Pierce Reserpine Standard used for?

The Thermo Scientific Pierce Reserpine Standard for LCMS is a standard for determining the performance and sensitivity of all Thermo Scientific mass spectrometry instruments. This standard must be diluted before use according to the procedure specified in the corresponding instrument's Getting Started guide.

Is there a way to evaluate the sensitivity of my triple quad mass spectrometry instrument?

Yes, Pierce Reserpine Standard for LC-MS (Cat. No. 88326) can be used to assess instrument sensitivity.

My spray stability is erratic in my mass spectrometer, can you help?

Yes, we offer the Pierce Reserpine Standard for LC-MS (Cat. No. 88326) to guide in troubleshooting spray stability.

For Research Use Only. Not for use in diagnostic procedures.