missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein Precipitation Plates

Catalog No. PI90036
Change view
Click to view available options
Quantity:
1 Kit (10 Plates)
1 Kit (2 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI90036 1 Kit (2 Plates)
PI90037 1 Kit (10 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI90036 Supplier Thermo Scientific™ Supplier No. 90036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pierce Protein Precipitation Plates are used for protein processing via simple, rapid and automatable protein precipitation and removal of proteins from plasma, serum and other biological fluid samples alongside the Rapid Equilibrium Device (RED) for plasma protein-binding assays.

Thermo Scientific Pierce Protein Precipitation Plates provide an easy, 96-well format for simple, rapid and automatable protein precipitation and filtration (purification) from plasma, serum and other biological fluid samples.

Pierce™ Protein Precipitation Plates are designed specifically for solvent-based precipitation of proteins and their subsequent removal from fluid samples. The plates are leak-free and retain organic solvent without dripping, even while mixing. The 2 mL well volume allows processing sample volumes of 15 to 400 μL per well, using the recommended precipitation conditions of 3:1 (v/v) solvent-to-serum ratio. The Protein Precipitation Plates are the ideal choice for downstream processing of plasma protein binding assay samples prepared with rapid equilibrium dialysis (RED) systems that are important for the pharmaceutical industry to perform plasma protein-binding assays, which comprise a critical step in drug development. Pierce Protein Precipitation Plates use a special graded filter system that retains precipitated protein particles without clogging, yet freely allows soluble (nonprecipitated) proteins to elute through the 0.2 μM cut-off membrane. This design maximizes protein removal and ensures the highest possible level of sample recovery.

Features of Protein Precipitation Plates:
• Prepare small molecule samples for HPLC or LC-MS in 10–30 minutes
• Compatible with most organic solvents
• No acetonitrile leakage for up to four hours
• Leach-free graded hydrophobic frit prevents blockage
• Filtrates are easily collected by vacuum, positive pressure or centrifugation
• Assay serum samples of 15 to 400 μL per well (2 mL well volume)

Specifications

For Use With (Equipment) Mass Spectrometer, Centrifuge
Product Type Protein Precipitation Plate
Product Line Pierce
Content And Storage Upon receipt store plates at room temperature. Product shipped at ambient temperature.
Starting Material Protease-digested Protein
Final Product Type Peptides
Detection Method Mass Spectrometry
Format Plate
Workflow Step Sample Clean Up
Quantity 1 Kit (2 Plates)