missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride)

Catalog No. PIPG82079
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPG82079 1 g
PIPG82073 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPG82079 Supplier Thermo Scientific™ Supplier No. PG82079
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize applications where product integrity and risk minimization are paramount with this high-quality formulation of carbodiimide crosslinker.

Thermo Scientific Pierce Premium Grade EDC is our highest quality formulation of this popular carbodiimide crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

Features of Premium Grade EDC:

  • EDC—carbodiimide crosslinker for conjugation between carboxylates and amines
  • High quality—identity and purity confirmed by several tests
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade EDC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. EDC is 1-ethyl-3-[3-dimethylaminopropyl]carbodiimide hydrochloride, a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation. In the presence of N-hydroxysulfosuccinimide (sulfo-NHS), EDC can be used to convert carboxyl groups to amine-reactive sulfo-NHS esters. This is accomplished by mixing the EDC with a carboxyl containing molecule and adding sulfo-NHS.

Specifications of Premium Grade EDC

We manufacture crosslinking reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

  • Visual: White crystalline powder, free of foreign matter
  • Identity: IR scan shows only peaks characteristic of EDC
  • Water content: Karl Fischer titration, ≤ 2%
  • Trace metals: All metals, including lead, ≤ 50 ppm

Applications:

  • Conjugate carboxyl and amino groups among peptides and proteins
  • Couple haptens to immunogenic carrier proteins (e.g., attach a peptide to KLH)
  • Immobilize peptide antigens to affinity-purify antibodies (Cat. No. 20266)
  • Create NHS-activated, amine-reactive labeling compounds
  • Crosslink proteins to carboxyl coated beads or surfaces
  • Activate nanoparticles with amine-reactive sulfo-NHS esters

Product References:

  • Crosslinker Application Guide -- search for recent literature references for this product

Related Products

  • EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride) (Cat. No. 22980)
  • Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide) (Cat. No. PG82071)

Specifications

Chemical Reactivity Amine-Carboxyl
Cleavable No
Molecular Weight (g/mol) 191.7
PEGylated No
Spacer Arm Length 0.0 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Room Temperature
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short (<10 Å)
Form Solid
Quantity 1 g
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Is your EDAC compound the same as your EDC compound?

Yes, both acronyms are used for the same compound. The chemical name for EDC is 1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide and the chemical name for EDAC is N-3-(3-dimethylaminopropyl)-N-ethyl-carbodiimide. Both of these compounds are structurally equivalent.

For Research Use Only. Not for use in diagnostic procedures.