missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PMPI (p-maleimidophenyl isocyanate)

Catalog No. PI28100
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28100 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI28100 Supplier Thermo Scientific™ Supplier No. LSG28100
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Short (8.7A) hydroxyl-to-sulfhydryl crosslinker; isocyanate and maleimide groups; label -OH by urethane bonds for conjugation to cysteines by thioether bonds.

Thermo Scientific™ Pierce PMPI is a maleimide-and-isocyanate crosslinker for attaching compounds to sulfhydryl groups (cysteines) after conjugating the linker to hydroxyl groups on a molecule by urethane (carbamate) bonds.

Highlights:

  • Maleimide forms stable thioether linkages
  • Isocyanate reacts with - OH groups to form a carbamate link at pH 8.5
  • Excellent tool for conjugating - OH group-containing compounds such as steroids and vitamins
  • Reactive groups: maleimide and isocyanate
  • Reactive toward: sulfhydryl and hydroxyl groups

Specifications

Chemical Reactivity Carboxyl-Sulfhydryl
Cleavable No
Description PMPI
Molecular Weight (g/mol) 307.1
PEGylated No
Spacer Arm Length 8.7 Å
Content And Storage Upon receipt store at 4°C protected from moisture.
Cell Permeability Not determined
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Isocyanate
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.