missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips

Catalog No. A40012259
Change view
Click to view available options
Sorbent:
MCX
SAX
SCX
SDB-RPS
SDB-XC
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent
A40012259 SDB-XC
A40012253 SDB-RPS
A40012258 SAX
A40012251 SCX
A40012256 MCX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A40012259 Supplier Thermo Scientific™ Supplier No. A40012259

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips are sorbent-based devices designed for rapid, reproducible peptide desalting, clean-up, and concentration prior to liquid chromatography-mass spectrometry (LC-MS) and other mass spectrometry (MS) workflows.

Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips are sorbent-based devices designed for rapid, reproducible peptide desalting, clean-up, and concentration prior to liquid chromatography-mass spectrometry (LC-MS) and other mass spectrometry (MS) workflows. These spin tips enable efficient peptide binding and recovery while removing salts, detergents, buffers, denaturants, chaotropes, and other chemistry-compatible contaminants that can interfere with downstream analysis. Available chemistries include octadecyl silica (C18), strong anion exchange (SAX), strong cation exchange (SCX), mixed-mode cation exchange (MCX), styrene divinylbenzene reversed-phase sulfonate (SDB-RPS), and styrene divinylbenzene crosslinked (SDB-XC), enabling selection based on the sample matrix, contaminant profile, and desired clean-up or fractionation strategy.

When used with Pierce™ Spin Tip adaptors (Catalog No. A40011776) and low protein binding microcentrifuge tubes (Catalog No. 90410), Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips support a simple centrifugation-based workflow with standard laboratory equipment. The spin tip format provides a straightforward setup for peptide desalting, clean-up, concentration, or chemistry-compatible fractionation before downstream MS analysis.

Features of Thermo Scientific™ Pierce™ Peptide Clean-Up Spin Tips

  • Efficient - help remove interfering contaminants to improve spectral quality and prepare peptide samples in mass spectrometry (MS)-compatible solutions
  • Flexible - choose from multiple chemistries to support peptide desalting, clean-up, concentration, and fractionation strategies
  • Convenient - process samples by standard centrifugation without specialized equipment using Pierce Spin Tip adaptors and low protein binding microcentrifuge tubes

Applications

  • Peptide desalting prior to liquid chromatography-mass spectrometry (LC-MS) analysis
  • Clean-up following enzymatic protein digestion workflows
  • Removal of chemistry-compatible salts, buffers, detergents, denaturants, chaotropes, lipids, and other liquid chromatography-mass spectrometry (LC-MS) interferences
  • Peptide concentration and mass spectrometry sample preparation
  • Peptide fractionation to reduce sample complexity and support deeper proteome analysis

Specifications

Product Type Spin Tip
Product Line Pierce
Starting Material Peptides, Protease-digested Protein
Detection Method Mass Spectrometry
Format Tip
Workflow Step Peptide Clean-up
Quantity 96 Tips
Form Solid
Sorbent SDB-XC

For Research Use Only. Not for use in diagnostic procedures.