missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PEG12-SPDP (PEGylated, long-chain SPDP crosslinker)

Catalog No. PI26129
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26129 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26129 Supplier Thermo Scientific™ Supplier No. 26129

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Very long (54.1A) amine-to-sulfhydryl crosslinker; NHS-ester and pyridyldithiol groups at ends of 12-unit polyethylene glycol spacer arm; forms cleavable disulfide bonds; water-soluble.

Thermo Scientific™ Pierce PEG12-SPDP is a multifunctional crosslinker for protein conjugation via amine-to-amine or amine-to-sulfhydryl crosslinks that contain a 12-unit polyethylene glycol (PEG) group and a reducible (cleavable) disulfide bond.

SPDP reagents have an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a suflhydryl-reactive 2-pyridyldithio group at the opposite end. PEG12-SPDP has a 12-unit polyethylene glycol spacer arm, which confers greater solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using an SPDP reagent.

Highlights:

  • Efficient amine conjugation – NHS ester provides effective conjugation to lysines, N-termini of peptides and other primary amines

  • Reversible sulfhydryl conjugation – pyridyldithio group enables disulfide linkage with reduced cysteines and other sulfhydryl groups, providing a stable but cleavable bond

  • Polyethylene glycol – PEG groups are flexible, non-immunogenic, hydrophilic, and often enhance the solubility of attached molecules

  • Spacer arm – connects primary amines and sulfhydryl targets at distances up to 54.1 angstroms

Specifications:

  • Chemical name (simplified): 2-pyridyldithiol-tetraoxaoctatriacontane-N-hydroxysuccinimide
  • Chemical formula: C39H65N3O17S2
  • Molecular weight: 912.07
  • Spacer arm: 54.1 angstroms
  • Form: light, viscous liquid
  • Solubility: dissolve 2mg in 110μL DMSO or DMF to make 20mM stock for each use

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description PEG12-SPDP
Molecular Weight (g/mol) 912.07
PEGylated Yes
Spacer Arm Length 54.1 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Pyridyldisulfide
Spacer Long (>30 Å)
Form Liquid
Quantity 100 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for PEG12-SPDP (PEGylated, long-chain SPDP crosslinker))?

PEG12-SPDP (PEGylated, long-chain SPDP crosslinker) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.