missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NHS-Phosphine

Catalog No. PI88900
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88900 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88900 Supplier Thermo Scientific™ Supplier No. 88900

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Derivatize proteins or amine surfaces for selective conjugation, detection or purification of azide-tagged targets with these activated phosphine compounds.

Thermo Scientific™ Pierce NHS-Phosphine and Sulfo-NHS-Phosphine are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to azide-tagged molecules.

These NHS-ester compounds react to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The phosphine groups (PPh2) react with azide-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples. In combination with different azide-tagged compounds, these NHS-Phosphine reagents facilitate a variety of labeling and crosslinking experimental strategies for investigating protein interactions and cellular pathways.

Highlights:

  • Versatile – Sulfo-NHS-Phosphine easily dissolves in water-miscible solvents for subsequent dilution in aqueous reaction mixtures for azide conjugation; NHS-Phosphine can be used for amine surface modification in organic solvents

  • Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents

  • Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)

  • Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency

Specifications

Chemical Reactivity Amine-Azide
Cleavable No
Description NHS-Phosphine
Molecular Weight (g/mol) 461.4
PEGylated No
Spacer Arm Length 5.4 Å
Content And Storage Upon receipt store at -20°C protected from light and moisture.
Cell Permeability Not determined
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety NHS Ester, Phosphine
Spacer Short (<10 Å)
Form Powder
Quantity 10 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for NHS-Phosphine?

NHS-Phosphine is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.