missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NHS-PEG4-Azide

Catalog No. PI26130
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26130 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26130 Supplier Thermo Scientific™ Supplier No. LSG26130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activated azido compounds to derivatize proteins or amine surfaces for selective conjugation, detection or purification by phosphine-modified probes.

Thermo Scientific™ Pierce NHS-Azide reagents are amine-reactive compounds that can be used to derivatize primary amines of proteins or amine-coated polymer surfaces for ligation to phosphine-modified molecules.

These NHS-ester compounds react to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples. In combination with different phosphine-activated compounds, these NHS-Azide reagents facilitate a variety of labeling and crosslinking experimental strategies for investigating protein interactions and cellular pathways.

Highlights:

  • Soluble – reagents easily dissolve in water-miscible solvents for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples

  • Compatible – reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents

  • Specific – NHS esters are specific for covalent attachment to primary amines (side chain of lysine or N-terminus of polypeptides)

  •  

  • Chemoselective – azide and phosphine groups do not react or interfere with components of biological samples but conjugate to one another with high efficiency

  • Pegylated – choose the very short NHS-Azide or one of the two varieties whose spacer arms contain medium (4-unit) or long (12-unit) polyethylene glycol (PEG) groups for increased solubility and reach

Specifications

Chemical Reactivity Amine-Phosphine
Cleavable No
Description NHS-PEG4-Azide
Molecular Weight (g/mol) 388.37
PEGylated Yes
Spacer Arm Length 18.9 Å
Content And Storage Upon receipt store at -20°C protected from light and moisture.
Cell Permeability Not determined
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Azide, NHS Ester
Spacer Medium (10 to 30 Å)
Form Liquid
Quantity 100 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for NHS-PEG4-Azide?

NHS-PEG4-Azide is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.