Learn More
Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kits
Achieve highly efficient, compatible immunoprecipitation and co-immunoprecipitation of target antigens for downstream LC-MS analysis using Thermo Scientific Pierce MS-Compatible Magnetic IP Kits with streptavidin or Protein A/G magnetic beads.
Supplier: Thermo Scientific™ LSG90408
Description
Immunoprecipitation is often necessary for the detection and quantitation of low-abundance proteins or rare signaling events. Mass spectrometry analysis relies on two methods for the detection of low-abundance proteins: single reaction monitoring (SRM) and parallel reaction monitoring (PRM), which enable isolation, scheduling, and quantitation of selected proteotypic peptides. These IP kits have been optimized for efficient immunoprecipitation and co-immunoprecipitation of target proteins, and are directly compatible with downstream MS sample preparation and analysis.
• MS-compatible—reagents directly compatible with in-solution peptide digestion• Sensitive—procedure successfully enriches low-abundance proteins (low ng range)
• Low background—enriched samples contain minimal detergent residuals
• Reduced antibody contamination—antibody biotinylation with streptavidin enrichment reduces contamination in the eluate when compared to other IP methods
• Robust—procedure and reagents have been verified with numerous targets to ensure consistent enrichment of low-abundance proteins (ng range) with at least two peptides identified per protein
The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone. The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads for use with biotinylated antibodies that may be purchased commercially or labeled using the Pierce Antibody Biotinylation Kit for IP. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion for LC-MS analysis without the need for gel purification, detergent removal, or desalting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.
Applications
• Enrichment of low-abundance targets to improve MS sensitivity
• Identification of target proteins that are not detectable by western blotting
• Quantitation of target proteins by LC-MS analysis
• Screening of antibodies for IP-MS
Specifications
| Streptavidin Magnetic Beads | |
| Magnetic IP Kit | |
| 40 Reactions | |
| Protein Samples, Cell Lysate | |
| Mass Spectrometry | |
| Sample Enrichment | |
| Mass Spectrometry |
| KingFisher™ Flex, Magnetic Stand | |
| Pierce | |
| Streptavidin Magnetic Beads, 1 mL IP-MS Cell Lysis Buffer, 100 mL IP-MS Wash Buffer A, 75 mL IP-MS Wash Buffer B, 40 mL IP-MS Elution Buffer, 6 mL Store in refrigerator (2° to 8°C). |
|
| Protein | |
| Kit | |
| 40 reactions kit |