missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MPBH (4-(4-N-maleimidophenyl)butyric acid hydrazide)

Catalog No. pi22305
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22305 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI22305 Supplier Thermo Scientific™ Supplier No. LSG22305

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this long (17.9A) crosslinker containing maleimide and hydrazide groups.

Thermo Scientific™ Pierce MPBH is a long, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Highlights:

  • Sulfhydryl-reactive maleimide forms a thioether linkage
  • Stable in DMSO
  • Reactive groups: hydrazide and maleimide
  • Reactive toward: carbohydrate and sulfhydryl groups
  • Use with EDC for reactivity toward carboxyl groups

Specifications

Chemical Reactivity Carbohydrate-Sulfhydryl
Cleavable No
Description MPBH
Molecular Weight (g/mol) 309.75
PEGylated No
Spacer Arm Length 17.9 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Amine, Hydrazide, Maleimide
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.